Learn More
Meso Scale Discovery U-PLEX Mouse IP-10 ASY 25 PL
SDP
Catalog No. 5000078441
Encompass
The Meso Scale Discovery U-PLEX Mouse IP-10 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology on patterned arrays for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and ancillary reagents needed to run the assay
Catalog No. 50-000-78441 Supplier Meso Scale Discovery Supplier No. K152UFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IP-10 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology on patterned arrays for high sensitivity and broad dynamic range The assay requires 50 L or less sample volume and can be read on the MESO QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and ancillary reagents needed to run the assay