Learn More
Meso Scale Discovery U-PLEX Mouse IP-10 ASY384-well
SDP
Catalog No. 5000081194
Encompass
The Meso Scale Discovery U-PLEX Mouse IP-10 (384-Well Assay) is a multiplex immunoassay kit for measuring mouse IP-10 in serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and calibrator solutions The detection range is 0 61 to 10 000 pg/mL requiring 25 L of sample with a total assay time of about 3 hours This product efficiently quantifies the chemokine IP-10
Catalog No. 50-000-81194 Supplier Meso Scale Discovery Supplier No. K252UFK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Mouse IP-10 (384-Well Assay) is a multiplex immunoassay kit for measuring mouse IP-10 in serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and rapid results The kit includes a 384-well plate with capture antibodies detection antibodies and calibrator solutions The detection range is 0 61 to 10 000 pg/mL requiring 25 L of sample with a total assay time of about 3 hours This product efficiently quantifies the chemokine IP-10