Learn More
Meso Scale Discovery U-PLEX Mouse IP-10 ASYQP
SDP
Catalog No. 5000079357
Encompass
The U-PLEX Mouse IP-10 Assay (K152UFK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure IP-10 (CXCL10) levels in mouse biological samples This single-plex assay utilizes Meso Scale Discovery s advanced U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The assay is optimized for various sample types including serum plasma and cell culture supernatants making it ideal for cytokine research applications in immunology and inflammation studies
Catalog No. 50-000-79357 Supplier Meso Scale Discovery Supplier No. K152UFK21
The U-PLEX Mouse IP-10 Assay (K152UFK) is a highly sensitive electrochemiluminescence immunoassay designed to quantitatively measure IP-10 (CXCL10) levels in mouse biological samples This single-plex assay utilizes Meso Scale Discovery s advanced U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The assay is optimized for various sample types including serum plasma and cell culture supernatants making it ideal for cytokine research applications in immunology and inflammation studies