Learn More
Meso Scale Discovery U-PLEX Mouse KC/GRO ASY
SDP
Catalog No. 5000079011
Encompass
The U-PLEX Mouse KC Assay K152VWK from Meso Scale Discovery is a multiplex immunoassay kit for measuring cytokines and chemokines in mouse samples It uses electrochemiluminescence detection to measure up to 10 analytes per well from a single sample The kit includes a 96-well plate detection antibodies calibrator solutions and buffers It requires 25 L of sample per well and can quantify IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IFN- TNF- and KC/GRO in various biological fluids
Catalog No. 50-000-79011 Supplier Meso Scale Discovery Supplier No. K152VWK2
The U-PLEX Mouse KC Assay K152VWK from Meso Scale Discovery is a multiplex immunoassay kit for measuring cytokines and chemokines in mouse samples It uses electrochemiluminescence detection to measure up to 10 analytes per well from a single sample The kit includes a 96-well plate detection antibodies calibrator solutions and buffers It requires 25 L of sample per well and can quantify IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IFN- TNF- and KC/GRO in various biological fluids