Learn More
Meso Scale Discovery U-PLEX Mouse Leptin ASY
SDP
Catalog No. 5000079382
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL for leptin
Catalog No. 50-000-79382 Supplier Meso Scale Discovery Supplier No. K1525ZK1
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls The kit has a sensitivity of 0 3 pg/mL and an upper limit of quantitation of 10 000 pg/mL for leptin