Learn More
Meso Scale Discovery U-PLEX Mouse Leptin ASYQP
SDP
Catalog No. 5000078447
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay for measuring mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours aiding in the quantitative determination of leptin levels in mouse samples
Catalog No. 50-000-78447 Supplier Meso Scale Discovery Supplier No. K1525ZK22
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay for measuring mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and precision The kit includes a plate with capture antibodies detection antibodies with electrochemiluminescent labels calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL It requires 25 L of sample and takes about 3 5 hours aiding in the quantitative determination of leptin levels in mouse samples