Learn More
Meso Scale Discovery U-PLEX Mouse Leptin ASYQP
SDP
Catalog No. 5000079987
Encompass
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit can perform up to 80 tests for leptin quantification
Catalog No. 50-000-79987 Supplier Meso Scale Discovery Supplier No. K1525ZK21
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse Leptin Assay Kit is a multiplex immunoassay that measures mouse leptin in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The assay requires 50 L or less sample volume and takes 3 5 hours to complete It includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls The kit can perform up to 80 tests for leptin quantification