Learn More
Meso Scale Discovery U-PLEX Mouse MCP-1 ASY384-well
SDP
Catalog No. 5000078954
Encompass
The Meso Scale Discovery Mouse MCP-1 Singleplex Assay is a sensitive immunoassay for measuring mouse monocyte chemoattractant protein-1 (MCP-1) in samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for accurate results with a detection limit of 0 3 pg/mL and a range up to 10 000 pg/mL The singleplex format allows MCP-1 measurement alone or with other biomarkers The kit includes all reagents and is compatible with MSD s SECTOR Imager platforms
Catalog No. 50-000-78954 Supplier Meso Scale Discovery Supplier No. K252UGK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Mouse MCP-1 Singleplex Assay is a sensitive immunoassay for measuring mouse monocyte chemoattractant protein-1 (MCP-1) in samples like serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection for accurate results with a detection limit of 0 3 pg/mL and a range up to 10 000 pg/mL The singleplex format allows MCP-1 measurement alone or with other biomarkers The kit includes all reagents and is compatible with MSD s SECTOR Imager platforms