Learn More
Meso Scale Discovery U-PLEX Mouse MCP-1 ASYQP
SDP
Catalog No. 5000078626
Encompass
MSD s U-PLEX Mouse MCP-1 assay is an electrochemiluminescence-based immunoassay for quantifying monocyte chemoattractant protein-1 in mouse samples This multiplex platform offers high sensitivity broad dynamic range and minimal sample volume requirements The assay utilizes proprietary SULFO-TAG labels and MULTI-SPOT plates for precise reproducible measurements of this key inflammatory biomarker in research applications
Catalog No. 50-000-78626 Supplier Meso Scale Discovery Supplier No. K152UGK22
MSD s U-PLEX Mouse MCP-1 assay is an electrochemiluminescence-based immunoassay for quantifying monocyte chemoattractant protein-1 in mouse samples This multiplex platform offers high sensitivity broad dynamic range and minimal sample volume requirements The assay utilizes proprietary SULFO-TAG labels and MULTI-SPOT plates for precise reproducible measurements of this key inflammatory biomarker in research applications