Learn More
Meso Scale Discovery U-PLEX Mouse MCP-5/CCL12 ASYQP
SDP
Catalog No. 5000080337
Encompass
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection for mouse MCP-5 is 1 3 pg/mL
Catalog No. 50-000-80337 Supplier Meso Scale Discovery Supplier No. K152Y1K22
The Meso Scale Discovery U-PLEX Mouse MCP-5 (CCL12) Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse MCP-5 in serum plasma and cell culture supernatants It utilizes electrochemiluminescent detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can perform up to 70 tests across two plates The lower limit of detection for mouse MCP-5 is 1 3 pg/mL