Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASY 25 PL
SDP
Catalog No. 5000082114
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex assay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines chemokines and growth factors (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 TNF- VEGF) from cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s ultra-sensitive MULTI-ARRAY technology and provides reagents for 1 000 wells enabling high-throughput cytokine profiling
Catalog No. 50-000-82114 Supplier Meso Scale Discovery Supplier No. K152X1K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex assay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines chemokines and growth factors (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 TNF- VEGF) from cell culture supernatants serum plasma and other biological samples This electrochemiluminescent assay utilizes MSD s ultra-sensitive MULTI-ARRAY technology and provides reagents for 1 000 wells enabling high-throughput cytokine profiling