Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASY
SDP
Catalog No. 5000081868
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring 10 mouse cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 TNF- ) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection and includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents Sensitivity ranges from 0 04-4 5 pg/mL enabling high-throughput sensitive quantification of inflammatory biomarkers
Catalog No. 50-000-81868 Supplier Meso Scale Discovery Supplier No. K152X1K1
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring 10 mouse cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 TNF- ) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection and includes a 96-well plate with capture antibodies detection antibodies calibrators and reagents Sensitivity ranges from 0 04-4 5 pg/mL enabling high-throughput sensitive quantification of inflammatory biomarkers