Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASY
SDP
Catalog No. 5000082004
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL5) in various samples It includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls The kit uses MSD s electrochemiluminescent technology for high sensitivity and minimal matrix interference in a multiplex format
Catalog No. 50-000-82004 Supplier Meso Scale Discovery Supplier No. K152X1K2
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 mouse cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF CXCL1 CCL2 CCL3 CCL5) in various samples It includes a 96-well plate with capture antibodies detection antibodies buffers diluent calibrators and controls The kit uses MSD s electrochemiluminescent technology for high sensitivity and minimal matrix interference in a multiplex format