Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASY384-well
SDP
Catalog No. 5000081153
Encompass
The U-PLEX Mouse MDC 384-Well Assay K252x1K from Meso Scale Diagnostics is a 384-well multiplex immunoassay kit for measuring mouse cytokines chemokines growth factors and other analytes in cell culture supernatants serum plasma and other biological matrices The kit contains reagents for measuring up to 252 analytes in a single sample using electrochemiluminescent detection technology The kit includes plates calibrators controls detection antibodies read buffer and other assay components required to complete a 384-well multiplex assay The kit enables high-throughput sensitive and reproducible quantification of multiple mouse proteins simultaneously using small sample volumes
Catalog No. 50-000-81153 Supplier Meso Scale Discovery Supplier No. K252X1K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse MDC 384-Well Assay K252x1K from Meso Scale Diagnostics is a 384-well multiplex immunoassay kit for measuring mouse cytokines chemokines growth factors and other analytes in cell culture supernatants serum plasma and other biological matrices The kit contains reagents for measuring up to 252 analytes in a single sample using electrochemiluminescent detection technology The kit includes plates calibrators controls detection antibodies read buffer and other assay components required to complete a 384-well multiplex assay The kit enables high-throughput sensitive and reproducible quantification of multiple mouse proteins simultaneously using small sample volumes