Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASYQP 25 PL
SDP
Catalog No. 5000082260
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines and chemokines (G-CSF GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 and TNF- ) from cell culture supernatants serum plasma and other biological samples This 10-plate kit provides reagents for over 1 000 assays in a quick and efficient electrochemiluminescent format compatible with MSD instrumentation
Catalog No. 50-000-82260 Supplier Meso Scale Discovery Supplier No. K152X1K24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines and chemokines (G-CSF GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 and TNF- ) from cell culture supernatants serum plasma and other biological samples This 10-plate kit provides reagents for over 1 000 assays in a quick and efficient electrochemiluminescent format compatible with MSD instrumentation