Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASYQP
SDP
Catalog No. 5000079060
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 CCL5 TNF- ) from cell culture supernatants serum plasma (EDTA heparin) and other biological samples This assay utilizes MSD s ultra-sensitive electrochemiluminescent detection technology and is provided in a convenient kit format that includes all necessary reagents and plates for high performance cytokine profiling
Catalog No. 50-000-79060 Supplier Meso Scale Discovery Supplier No. K152X1K21
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit that enables simultaneous quantitative measurement of 15 mouse myeloid-derived cytokines and chemokines (GM-CSF IFN- IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 CXCL1 CCL2 CCL3 CCL4 CCL5 TNF- ) from cell culture supernatants serum plasma (EDTA heparin) and other biological samples This assay utilizes MSD s ultra-sensitive electrochemiluminescent detection technology and is provided in a convenient kit format that includes all necessary reagents and plates for high performance cytokine profiling