Learn More
Meso Scale Discovery U-PLEX Mouse MDC ASYQP
SDP
Catalog No. 5000082113
Encompass
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 mouse myeloid cell markers (CD11b CD11c CD14 CD16 CD19 CD45 CD64 CD86 CD115 CD124 CD172a F4/80 Ly6C Ly6G MHC-II) in cell lysates serum plasma and other samples It uses MSD s electrochemiluminescent detection offering high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture and detection antibodies calibrators and controls compatible with MSD QuickPlex SQ120 and SECTOR Imager readers
Catalog No. 50-000-82113 Supplier Meso Scale Discovery Supplier No. K152X1K22
The U-PLEX Mouse MDC Assay K152x1K from Meso Scale Discovery is a multiplex immunoassay kit for measuring 15 mouse myeloid cell markers (CD11b CD11c CD14 CD16 CD19 CD45 CD64 CD86 CD115 CD124 CD172a F4/80 Ly6C Ly6G MHC-II) in cell lysates serum plasma and other samples It uses MSD s electrochemiluminescent detection offering high sensitivity a 5-log dynamic range and low sample volume The kit includes a 96-well plate with capture and detection antibodies calibrators and controls compatible with MSD QuickPlex SQ120 and SECTOR Imager readers