Learn More
Meso Scale Discovery U-PLEX Mouse MIP-1 ASY
SDP
Catalog No. 5000078472
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha Assay Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 3 2 pg/mL to 100 000 pg/mL
Catalog No. 50-000-78472 Supplier Meso Scale Discovery Supplier No. K152UJK2
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha Assay Kit is a multiplex immunoassay that quantitatively measures mouse MIP-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on Meso Scale Discovery SECTOR Imagers The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 3 2 pg/mL to 100 000 pg/mL