Learn More
Meso Scale Discovery U-PLEX Mouse MIP-1 ASY
SDP
Catalog No. 5000080450
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha Assay Kit is a multiplex immunoassay that measures mouse macrophage inflammatory protein 1-alpha (MIP-1alpha) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to run and 10 L of sample is required The kit can test up to 80 samples in duplicate
Catalog No. 50-000-80450 Supplier Meso Scale Discovery Supplier No. K152UJK1
The Meso Scale Discovery U-PLEX Mouse MIP-1alpha Assay Kit is a multiplex immunoassay that measures mouse macrophage inflammatory protein 1-alpha (MIP-1alpha) in serum plasma and cell culture supernatants The assay uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and diluent The assay takes approximately 3 5 hours to run and 10 L of sample is required The kit can test up to 80 samples in duplicate