Learn More
Meso Scale Discovery U-PLEX Mouse MIP-1 ASYQP
SDP
Catalog No. 5000079136
Encompass
The U-PLEX Mouse MIP-1 384-Well Assay (K252UJK) is a highly sensitive immunoassay designed for quantitative measurement of mouse macrophage inflammatory protein-1 alpha in biological samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The 384-well format enables high-throughput screening capabilities for research applications studying inflammation immune responses and cytokine signaling pathways in mouse models
Catalog No. 50-000-79136 Supplier Meso Scale Discovery Supplier No. K152UJK21
The U-PLEX Mouse MIP-1 384-Well Assay (K252UJK) is a highly sensitive immunoassay designed for quantitative measurement of mouse macrophage inflammatory protein-1 alpha in biological samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering superior sensitivity and dynamic range compared to traditional ELISA methods The 384-well format enables high-throughput screening capabilities for research applications studying inflammation immune responses and cytokine signaling pathways in mouse models