Learn More
Meso Scale Discovery U-PLEX Mouse MIP-1β Assay, 5 plates
SDP
Catalog No. NC1668063
Encompass
The U-PLEX Mouse MIP-1β Assay maximizes your flexibility to design an experiment that measures MIP-1β alone or multiplexed with other MSD U-PLEX assays, your own antibody pairs, or a combination of both. This assay accurately quantitates native and recombinant mouse MIP-1β in multiple sample types.
Catalog No. NC1668063 Supplier Meso Scale Discovery Supplier No. K152UKK2
The U-PLEX Mouse MIP-1β Assay maximizes your flexibility to design an experiment that measures MIP-1β alone or multiplexed with other MSD U-PLEX assays, your own antibody pairs, or a combination of both. This assay accurately quantitates native and recombinant mouse MIP-1β in multiple sample types.