Learn More
Meso Scale Discovery U-PLEX Mouse MIP-1B ASYQP
SDP
Catalog No. 5000078445
Encompass
MSD offers U-PLEX mouse MIP-1 (CCL4) immunoassays for quantitative detection of this chemokine biomarker The electrochemiluminescence-based platform provides sensitive specific measurement of MIP-1 levels in mouse samples U-PLEX technology enables multiplexed analysis with minimal sample volume requirements and wide dynamic range for research applications studying inflammation immune responses and disease pathways in mouse models
Catalog No. 50-000-78445 Supplier Meso Scale Discovery Supplier No. K152UKK21
MSD offers U-PLEX mouse MIP-1 (CCL4) immunoassays for quantitative detection of this chemokine biomarker The electrochemiluminescence-based platform provides sensitive specific measurement of MIP-1 levels in mouse samples U-PLEX technology enables multiplexed analysis with minimal sample volume requirements and wide dynamic range for research applications studying inflammation immune responses and disease pathways in mouse models