Learn More
Meso Scale Discovery U-PLEX Mouse MIP-2 ASY
SDP
Catalog No. 5000080341
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-2 Assay is a multiplex immunoassay kit for measuring mouse MIP-2 in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents Results are available in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 providing a rapid sensitive and cost-effective method to profile MIP-2 levels
Catalog No. 50-000-80341 Supplier Meso Scale Discovery Supplier No. K152XLK1
The Meso Scale Discovery U-PLEX Mouse MIP-2 Assay is a multiplex immunoassay kit for measuring mouse MIP-2 in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity and a wide dynamic range The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents Results are available in 3 5 hours using an MSD instrument like MESO QuickPlex SQ 120 providing a rapid sensitive and cost-effective method to profile MIP-2 levels