Learn More
Meso Scale Discovery U-PLEX Mouse MIP-2 ASYQP
SDP
Catalog No. 5000082115
Encompass
MSD s U-PLEX Mouse MIP-2 assay is an electrochemiluminescence-based immunoassay for quantifying macrophage inflammatory protein-2 (MIP-2) levels in mouse samples This multiplex-compatible assay utilizes MSD s proprietary SPOT technology for sensitive and specific detection of this chemokine biomarker The U-PLEX format allows researchers to customize panels by combining multiple analytes in a single well improving efficiency and sample conservation Designed for research applications studying inflammation immune responses and related pathways in mouse models
Catalog No. 50-000-82115 Supplier Meso Scale Discovery Supplier No. K152XLK22
MSD s U-PLEX Mouse MIP-2 assay is an electrochemiluminescence-based immunoassay for quantifying macrophage inflammatory protein-2 (MIP-2) levels in mouse samples This multiplex-compatible assay utilizes MSD s proprietary SPOT technology for sensitive and specific detection of this chemokine biomarker The U-PLEX format allows researchers to customize panels by combining multiple analytes in a single well improving efficiency and sample conservation Designed for research applications studying inflammation immune responses and related pathways in mouse models