Learn More
Meso Scale Discovery U-PLEX Mouse MIP-3 ASY
SDP
Catalog No. 5000078369
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha Assay is a multiplex immunoassay kit for measuring mouse MIP-3alpha in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents Results are analyzed on MSD s SECTOR Imager enabling rapid and accurate MIP-3alpha quantification important for immune function studies
Catalog No. 50-000-78369 Supplier Meso Scale Discovery Supplier No. K152UZK1
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha Assay is a multiplex immunoassay kit for measuring mouse MIP-3alpha in various samples It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents Results are analyzed on MSD s SECTOR Imager enabling rapid and accurate MIP-3alpha quantification important for immune function studies