Learn More
Meso Scale Discovery U-PLEX Mouse MIP-3 ASY
SDP
Catalog No. 5000082178
Encompass
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse macrophage inflammatory protein 3 alpha (MIP-3alpha) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 10-point standard curve for MIP-3alpha ranging from 2500 to 2 4 pg/mL
Catalog No. 50-000-82178 Supplier Meso Scale Discovery Supplier No. K152UZK2
The Meso Scale Discovery U-PLEX Mouse MIP-3alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse macrophage inflammatory protein 3 alpha (MIP-3alpha) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrator diluent wash buffer read buffer and a 10-point standard curve for MIP-3alpha ranging from 2500 to 2 4 pg/mL