Learn More
Meso Scale Discovery U-PLEX Mouse MMP-9 total ASY
SDP
Catalog No. 5000081340
Encompass
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is designed to quantitatively measure total MMP-9 (pro and active forms) in mouse biological samples such as serum plasma cell culture supernatants lavage fluids and tissue homogenates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide rapid and accurate results with high sensitivity and a broad dynamic range The assay requires a low sample volume of 50 L and has a lower limit of detection of 0 04 ng/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent
Catalog No. 50-000-81340 Supplier Meso Scale Discovery Supplier No. K152ZGK1
The Meso Scale Discovery U-PLEX Mouse MMP-9 Total Assay Kit is designed to quantitatively measure total MMP-9 (pro and active forms) in mouse biological samples such as serum plasma cell culture supernatants lavage fluids and tissue homogenates This electrochemiluminescent immunoassay utilizes MSD s proprietary SULFO-TAG labels and multi-array/multi-spot plate technology to provide rapid and accurate results with high sensitivity and a broad dynamic range The assay requires a low sample volume of 50 L and has a lower limit of detection of 0 04 ng/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent