Learn More
Meso Scale Discovery U-PLEX Mouse NGAL/LCN2 ASYQP
SDP
Catalog No. 5000079680
Encompass
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in mouse biological samples such as serum plasma and tissue culture supernatant This assay uses MSD s proprietary SECTOR Imager 6000 for detection and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be multiplexed with other U-PLEX assays to analyze multiple biomarkers simultaneously while conserving precious sample This product could be useful for studying kidney injury and other disease states involving NGAL
Catalog No. 50-000-79680 Supplier Meso Scale Discovery Supplier No. K152Z1K21
The Meso Scale Discovery U-PLEX Mouse NGAL (LCN2) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of Neutrophil Gelatinase-Associated Lipocalin (NGAL) in mouse biological samples such as serum plasma and tissue culture supernatant This assay uses MSD s proprietary SECTOR Imager 6000 for detection and provides high sensitivity broad dynamic range and rapid results The assay requires 50 L or less sample volume and can be multiplexed with other U-PLEX assays to analyze multiple biomarkers simultaneously while conserving precious sample This product could be useful for studying kidney injury and other disease states involving NGAL