Learn More
Meso Scale Discovery U-PLEX Mouse PYY total ASY
SDP
Catalog No. 5000082353
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for measuring total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective assay with just 25 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL ideal for high-throughput mouse research
Catalog No. 50-000-82353 Supplier Meso Scale Discovery Supplier No. K1526BK2
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay Kit is an electrochemiluminescent immunoassay for measuring total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for a rapid sensitive and cost-effective assay with just 25 L of sample The kit includes a 96-well plate with capture antibodies detection antibodies with SULFO-TAG calibrators and controls Sensitivity is 0 3 pg/mL with a quantification limit of 10 000 pg/mL ideal for high-throughput mouse research