Learn More
Meso Scale Discovery U-PLEX Mouse PYY total ASYQP
SDP
Catalog No. 5000082174
Encompass
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay K1526BK is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of total PYY levels down to 0 3 pg/mL The assay requires 50 L of sample and can be read on MSD s SECTOR Imager instruments This assay provides a simple rapid high-throughput method for measuring total PYY levels in mouse biological samples
Catalog No. 50-000-82174 Supplier Meso Scale Discovery Supplier No. K1526BK22
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX Mouse PYY Total Assay K1526BK is an electrochemiluminescent immunoassay for the quantitative measurement of total Peptide YY (PYY) in mouse serum plasma and cell culture supernatants This assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and accurate detection of total PYY levels down to 0 3 pg/mL The assay requires 50 L of sample and can be read on MSD s SECTOR Imager instruments This assay provides a simple rapid high-throughput method for measuring total PYY levels in mouse biological samples