Learn More
Meso Scale Discovery U-PLEX Mouse SDF-1 ASY
SDP
Catalog No. 5000078986
Encompass
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a quick 2 5 hour turnaround time per 96-well plate This assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate including a pre-coated plate detection antibody read buffer calibrator and controls
Catalog No. 50-000-78986 Supplier Meso Scale Discovery Supplier No. K152VBK1
The Meso Scale Discovery U-PLEX Mouse SDF-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantification of mouse SDF-1alpha in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision with a quick 2 5 hour turnaround time per 96-well plate This assay requires a Meso Scale Discovery SECTOR Imager and provides components for one 96-well plate including a pre-coated plate detection antibody read buffer calibrator and controls