Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery U-PLEX Mouse TARC Assay, Single-analyte, 96-well, EDTA Plasma/Serum, LLOD 0.32 pg/mL, Dynamic Range 0.32 - 1,200 pg/mL, Capture Antibody Goat Polyclonal, Detection Antibody Goat Polyclonal
SDP

Catalog No. NC3094908
Encompass

The MESO SCALE DISCOVERY U-PLEX Mouse TARC Assay is a sensitive and robust immunoassay for measuring the Thymus and activation-regulated chemokine (TARC) in mouse samples. Utilizing the U-PLEX platform, this assay features a 96-well format that is ideal for quantitative research applications in fields such as immunology and inflammation.

  • Platform: U-PLEX
  • Analyte: TARC (Thymus and activation-regulated chemokine, Ccl17)
  • Species reactivity: Mouse
  • Assay format: Single-analyte
  • Plate type: 96-well
  • Application: Cytokines & Chemokines, Immunology/Inflammation
  • Lower limit of detection (LLOD): 0.32 pg/mL
  • Dynamic range: 0.32 - 1,200 pg/mL
  • Sample type: EDTA Plasma, Serum
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM, MESO QuickPlex Q 60MM, and others
  • Research use only: Yes
  • Storage condition: Multi-Component, refer to product insert

Catalog No. NC3094908 Supplier Meso Scale Discovery Supplier No. K152B0K2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY U-PLEX Mouse TARC Assay is a sensitive and robust immunoassay for measuring the Thymus and activation-regulated chemokine (TARC) in mouse samples. Utilizing the U-PLEX platform, this assay features a 96-well format that is ideal for quantitative research applications in fields such as immunology and inflammation.

  • Platform: U-PLEX
  • Analyte: TARC (Thymus and activation-regulated chemokine, Ccl17)
  • Species reactivity: Mouse
  • Assay format: Single-analyte
  • Plate type: 96-well
  • Application: Cytokines & Chemokines, Immunology/Inflammation
  • Lower limit of detection (LLOD): 0.32 pg/mL
  • Dynamic range: 0.32 - 1,200 pg/mL
  • Sample type: EDTA Plasma, Serum
  • Compatible instruments: MESO SECTOR S 600MM, MESO QuickPlex SQ 120MM, MESO QuickPlex Q 60MM, and others
  • Research use only: Yes
  • Storage condition: Multi-Component, refer to product insert

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.