Learn More
Meso Scale Discovery U-PLEX Mouse TARC ASY
SDP
Catalog No. 5000081353
Encompass
The U-PLEX Mouse TARC Assay from Meso Scale Discovery is a multiplex immunoassay measuring TARC in mouse samples It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less it works with serum plasma cell culture supernatants and lavage fluids The kit includes reagents for one 96-well plate testing 40 samples in duplicate Detection limits are 0 3 pg/mL to 10 000 pg/mL for TARC
Catalog No. 50-000-81353 Supplier Meso Scale Discovery Supplier No. K152B0K1
Add to Cart
Add to Cart
The U-PLEX Mouse TARC Assay from Meso Scale Discovery is a multiplex immunoassay measuring TARC in mouse samples It uses electrochemiluminescence detection for high sensitivity broad dynamic range and precision Requiring 50 L or less it works with serum plasma cell culture supernatants and lavage fluids The kit includes reagents for one 96-well plate testing 40 samples in duplicate Detection limits are 0 3 pg/mL to 10 000 pg/mL for TARC