Learn More
Meso Scale Discovery U-PLEX Mouse TARC ASY384-well
SDP
Catalog No. 5000079980
Encompass
The U-PLEX Mouse TARC 384-well Assay K252B0K from Meso Scale Discovery is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Thymus and Activation Regulated Chemokine (TARC/CCL17) in mouse serum plasma and cell culture supernatants This 10-plex assay has a detection range of 0 03-2500 pg/mL and includes reagents for one 384-well plate The assay utilizes MSD GOLD technology provides high sensitivity and has a rapid 2 5 hour protocol This assay is for research use only
Catalog No. 50-000-79980 Supplier Meso Scale Discovery Supplier No. K252B0K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Mouse TARC 384-well Assay K252B0K from Meso Scale Discovery is a 384-well electrochemiluminescent immunoassay for the quantitative measurement of Thymus and Activation Regulated Chemokine (TARC/CCL17) in mouse serum plasma and cell culture supernatants This 10-plex assay has a detection range of 0 03-2500 pg/mL and includes reagents for one 384-well plate The assay utilizes MSD GOLD technology provides high sensitivity and has a rapid 2 5 hour protocol This assay is for research use only