Learn More
Meso Scale Discovery U-PLEX Mouse TGF-β1 Assay, 1 plate
SDP
Catalog No. NC3237678
Encompass
The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. The U-PLEX Mouse TGF-β1 Assay has the essential components for an assay that measures TGF-β1 in a single-analyte format. For information on multiplexing TGF-β1 with other analytes, please see U-PLEX Custom Biomarker Group 2 (mouse) Assays.
Catalog No. NC3237678 Supplier Meso Scale Discovery Supplier No. K152XWK1
The U-PLEX platform offers validated components that can be combined to create high quality assays quickly and easily. The U-PLEX Mouse TGF-β1 Assay has the essential components for an assay that measures TGF-β1 in a single-analyte format. For information on multiplexing TGF-β1 with other analytes, please see U-PLEX Custom Biomarker Group 2 (mouse) Assays.