Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B1 ASYQP
SDP
Catalog No. 5000078695
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta1 with a lower limit of detection of 1 7 pg/mL and an upper limit of 100 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and reagents sufficient for one 96-well plate Sample incubation time is just 2 hours
Catalog No. 50-000-78695 Supplier Meso Scale Discovery Supplier No. K152XWK22
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta1 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta1 with a lower limit of detection of 1 7 pg/mL and an upper limit of 100 000 pg/mL The kit includes a 96-well plate coated with capture antibody detection antibody calibrator diluent wash buffer read buffer and reagents sufficient for one 96-well plate Sample incubation time is just 2 hours