Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B1 ASYQP
SDP
Catalog No. 5000082415
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active and total TGF-beta1 in mouse serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SECTOR Imager and provides detection limits down to 0 3 pg/mL with an assay time of 3 5 hours This multiplex assay allows simultaneous measurement of TGF-beta1 along with other biomarkers It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance
Catalog No. 50-000-82415 Supplier Meso Scale Discovery Supplier No. K152XWK21
The Meso Scale Discovery U-PLEX Mouse TGF-beta1 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active and total TGF-beta1 in mouse serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SECTOR Imager and provides detection limits down to 0 3 pg/mL with an assay time of 3 5 hours This multiplex assay allows simultaneous measurement of TGF-beta1 along with other biomarkers It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance