Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B2 ASY
SDP
Catalog No. 5000081871
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This product can aid in the quantitative determination of TGF-beta 2 levels in mouse samples
Catalog No. 50-000-81871 Supplier Meso Scale Discovery Supplier No. K152XUK1
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and assay diluent The assay has a lower limit of detection of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL The total assay time is 3 5 hours This product can aid in the quantitative determination of TGF-beta 2 levels in mouse samples