Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B2 ASY
SDP
Catalog No. 5000082413
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 1 3 pg/mL This product enables sensitive reproducible quantification of mouse TGF-beta 2 for research applications
Catalog No. 50-000-82413 Supplier Meso Scale Discovery Supplier No. K152XUK2
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides high sensitivity and a broad dynamic range The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent The assay takes approximately 3 5 hours to complete and has a lower limit of detection of 1 3 pg/mL This product enables sensitive reproducible quantification of mouse TGF-beta 2 for research applications