Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B2 ASYQP
SDP
Catalog No. 5000081336
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL
Catalog No. 50-000-81336 Supplier Meso Scale Discovery Supplier No. K152XUK21
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of mouse TGF-beta 2 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to MSD SULFO-TAG calibrators and sample diluent The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL