Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B2 ASYQP
SDP
Catalog No. 5000082414
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for measuring TGF-beta 2 in mouse serum plasma and cell culture supernatants It uses MSD s SECTOR Imager for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator Sensitivity is 0 3 pg/mL with an upper limit of 10 000 pg/mL The assay time is 3 5 hours enabling rapid sensitive quantification of TGF-beta 2 from mouse samples
Catalog No. 50-000-82414 Supplier Meso Scale Discovery Supplier No. K152XUK22
The Meso Scale Discovery U-PLEX Mouse TGF-beta 2 Assay Kit is an electrochemiluminescent immunoassay for measuring TGF-beta 2 in mouse serum plasma and cell culture supernatants It uses MSD s SECTOR Imager for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody Read Buffer Calibrator Diluent and lyophilized U-PLEX Calibrator Sensitivity is 0 3 pg/mL with an upper limit of 10 000 pg/mL The assay time is 3 5 hours enabling rapid sensitive quantification of TGF-beta 2 from mouse samples