Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B3 ASY
SDP
Catalog No. 5000079110
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for high-throughput quantification of TGF-beta3 levels
Catalog No. 50-000-79110 Supplier Meso Scale Discovery Supplier No. K152XVK2
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager and provides a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for high-throughput quantification of TGF-beta3 levels