Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B3 ASY
SDP
Catalog No. 5000081998
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse TGF-beta3 in serum plasma cell culture supernatants and other samples It uses the QuickPlex SQ 120 instrument and multi-array technology for sensitive detection in 25 L samples The kit includes a 96-well plate with capture antibodies detection antibodies calibrators diluent wash buffer read buffer and seals Sensitivity is 1 3 pg/mL with an upper quantification limit of 10 000 pg/mL
Catalog No. 50-000-81998 Supplier Meso Scale Discovery Supplier No. K152XVK1
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for quantifying mouse TGF-beta3 in serum plasma cell culture supernatants and other samples It uses the QuickPlex SQ 120 instrument and multi-array technology for sensitive detection in 25 L samples The kit includes a 96-well plate with capture antibodies detection antibodies calibrators diluent wash buffer read buffer and seals Sensitivity is 1 3 pg/mL with an upper quantification limit of 10 000 pg/mL