Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B3 ASYQP
SDP
Catalog No. 5000079647
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for the accurate quantification of TGF-beta3 levels in mouse biological samples
Catalog No. 50-000-79647 Supplier Meso Scale Discovery Supplier No. K152XVK21
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of TGF-beta3 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary SECTOR Imager to provide a sensitive and rapid method for measuring TGF-beta3 with a detection range of 1 5-1000 pg/mL and an assay time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody calibrator and sample diluent This assay allows for the accurate quantification of TGF-beta3 levels in mouse biological samples