Learn More
Meso Scale Discovery U-PLEX Mouse TGF-B3 ASYQP
SDP
Catalog No. 5000080336
Encompass
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit measures TGF-beta3 in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels for sensitive quantification down to sub-pg/mL levels It includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent The assay takes about 3 5 hours and suits lab automation It enables high-throughput analysis of TGF-beta3 a key cytokine in development and disease
Catalog No. 50-000-80336 Supplier Meso Scale Discovery Supplier No. K152XVK22
The Meso Scale Discovery U-PLEX Mouse TGF-beta3 Assay Kit measures TGF-beta3 in mouse samples like serum plasma and tissue lysates This electrochemiluminescent immunoassay uses MSD s SULFO-TAG labels for sensitive quantification down to sub-pg/mL levels It includes a 96-well plate with capture antibodies detection antibodies calibrators and diluent The assay takes about 3 5 hours and suits lab automation It enables high-throughput analysis of TGF-beta3 a key cytokine in development and disease