Learn More
Meso Scale Discovery U-PLEX Mouse TNF- ASY
SDP
Catalog No. 5000079352
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF-alpha Assay is a multiplex immunoassay kit that enables simultaneous measurement of mouse tumor necrosis factor alpha (TNF-alpha) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The lower limit of detection is 0 04 pg/mL
Catalog No. 50-000-79352 Supplier Meso Scale Discovery Supplier No. K152UCK2
The Meso Scale Discovery U-PLEX Mouse TNF-alpha Assay is a multiplex immunoassay kit that enables simultaneous measurement of mouse tumor necrosis factor alpha (TNF-alpha) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The lower limit of detection is 0 04 pg/mL