Learn More
Meso Scale Discovery U-PLEX Mouse TNF-RI ASY
SDP
Catalog No. 5000078934
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer for sample dilution It can accurately quantify TNFR1 levels with minimal sample volume requirements This kit is for research use only
Catalog No. 50-000-78934 Supplier Meso Scale Discovery Supplier No. K1520VK1
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse tumor necrosis factor receptor 1 (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer for sample dilution It can accurately quantify TNFR1 levels with minimal sample volume requirements This kit is for research use only