Learn More
Meso Scale Discovery U-PLEX Mouse TNF-RI ASY
SDP
Catalog No. 5000081849
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse tumor necrosis factor receptor I (TNF RI) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer for sample dilution It can test up to 80 samples in duplicate and has a testing time of 3 5 hours The assay has high specificity and shows minimal cross-reactivity
Catalog No. 50-000-81849 Supplier Meso Scale Discovery Supplier No. K1520VK2
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse tumor necrosis factor receptor I (TNF RI) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer for sample dilution It can test up to 80 samples in duplicate and has a testing time of 3 5 hours The assay has high specificity and shows minimal cross-reactivity