Learn More
Meso Scale Discovery U-PLEX Mouse TNF-RI ASYQP
SDP
Catalog No. 5000079871
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay for measuring mouse TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers It quantifies TNFR1 levels from 100 pg/mL to 10 000 pg/mL with minimal sample volume enabling reliable quantification of soluble TNFR1 crucial in TNF signaling
Catalog No. 50-000-79871 Supplier Meso Scale Discovery Supplier No. K1520VK21
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay for measuring mouse TNF receptor I (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection for high sensitivity broad dynamic range and precision The kit includes a 96-well plate with capture antibodies ECL-tagged detection antibodies calibrators and buffers It quantifies TNFR1 levels from 100 pg/mL to 10 000 pg/mL with minimal sample volume enabling reliable quantification of soluble TNFR1 crucial in TNF signaling