Learn More
Meso Scale Discovery U-PLEX Mouse TNF-RI ASYQP
SDP
Catalog No. 5000079872
Encompass
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse TNF receptor 1 (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer reagents needed to perform the assay It can detect mouse TNFR1 over a range of 78 pg/mL to 100 000 pg/mL
Catalog No. 50-000-79872 Supplier Meso Scale Discovery Supplier No. K1520VK22
The Meso Scale Discovery U-PLEX Mouse TNF RI Assay Kit is a multiplex immunoassay that measures mouse TNF receptor 1 (TNFR1) in serum plasma and cell culture supernatants It uses electrochemiluminescence (ECL) detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an ECL tag calibrators and buffer reagents needed to perform the assay It can detect mouse TNFR1 over a range of 78 pg/mL to 100 000 pg/mL